Anti-DNA Mouse Monoclonal Antibody [clone: 5F151]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3875-01EA 4430 DKK
USBID3875-01
Anti-DNA Mouse Monoclonal Antibody [clone: 5F151]
Antistoffer
Recommended dilution: Immunochemistry: 1:10

Type: Primary
Antigen: DNA
Clonality: monoclonal
Clone: 5F151
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Xenopus laevis
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR