Anti-Dkk-4 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3810-06EA 3070 DKK
USBID3810-06
Anti-Dkk-4 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 0.5-4 ?g/ml

Type: Primary
Antigen: Dkk-4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR