Anti-Dkk-3 Mouse monoclonal antibody [clone: 7H46]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3810-05EA 7150 DKK
USBID3810-05
Anti-Dkk-3 Mouse monoclonal antibody [clone: 7H46]
Antistoffer
Recommended dilution: Western Blot: 1-2?g/ml

Type: Primary
Antigen: Dkk-3
Clonality: monoclonal
Clone: 7H46
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR