Anti-Dkk-2 CT Rabbit Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3810-03B-BIOTEA 4620 DKK
USBID3810-03B-BIOT
Anti-Dkk-2 CT Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: Dkk-2 CT
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR