Anti-Dkk-2 CT Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3810-03BEA 4380 DKK
USBID3810-03B
Anti-Dkk-2 CT Rabbit polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: Dkk-2 CT
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR