Anti-Dkk-2 Rabbit Polyclonal Antibody (Biotin)
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBID3810-03A-BIOTEA 6360 DKK
USBID3810-03A-BIOT
Anti-Dkk-2 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: Dkk-2
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human