Anti-Dkk-2 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3810-03EA 3350 DKK
USBID3810-03
Anti-Dkk-2 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: Dkk-2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR