Anti-DKK1 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3810-01MEA 3310 DKK
USBID3810-01M
Anti-DKK1 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 0.5-2?g/ml

Type: Primary
Antigen: DKK1 (dickkopf WNT signaling pathway inhibitor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR