Anti-DLX2 Mouse Monoclonal Antibody [clone: 2E12]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3800-04EEA 4790 DKK
USBID3800-04E
Anti-DLX2 Mouse Monoclonal Antibody [clone: 2E12]
Antistoffer
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: DLX2 (distal-less homeobox 2)
Clonality: monoclonal
Clone: 2000000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR