Anti-DLX2 Mouse Monoclonal Antibody [clone: 4B9]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3800-04CEA 4730 DKK
USBID3800-04C
Anti-DLX2 Mouse Monoclonal Antibody [clone: 4B9]
Antistoffer
Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: DLX2 (distal-less homeobox 2)
Clonality: monoclonal
Clone: 4B9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR