Anti-DLX4 Mouse Monoclonal Antibody [clone: 8J108]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3800-04BEA 3030 DKK
USBID3800-04B
Anti-DLX4 Mouse Monoclonal Antibody [clone: 8J108]
Antistoffer
Recommended dilution: Immunochemistry: 2-100 ?g/ml

Type: Primary
Antigen: DLX4 (distal-less homeobox 4)
Clonality: monoclonal
Clone: 8J108
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR