Anti-DLX3 Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2728]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3800-03-APEA 0 DKK
USBID3800-03-AP
Anti-DLX3 Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2728]
Type: Primary
Antigen: DLX3 (distal-less homeobox 3)
Clonality: monoclonal
Clone: 10B2728
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR