Anti-DLGAP4 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3799-18EA 5140 DKK
USBID3799-18
Anti-DLGAP4 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: DLGAP4 (discs, large (Drosophila) homolog-associated protein 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR