Anti-DKK2 Goat polyclonal antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBID3795-03CEA 4120 DKK
USBID3795-03C
Anti-DKK2 Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1: 8000; Western Blot: 1-3?g/ml
Type: Primary
Antigen: DKK2 (dickkopf WNT signaling pathway inhibitor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human