Anti-DKK3 Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3795-02DEA 3460 DKK
USBID3795-02D
Anti-DKK3 Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:16000; Western Blot: 0. 03-0.1 ?g/ml

Type: Primary
Antigen: DKK3 (dickkopf WNT signaling pathway inhibitor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR