Anti-DKC1 Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3790-15CEA 3460 DKK
USBID3790-15C
Anti-DKC1 Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:128000; Immunochemistry: 5-10 ?g/ml

Type: Primary
Antigen: DKC1 (dyskeratosis congenita 1, dyskerin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR