Anti-Dixin Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3785EA 3460 DKK
USBID3785
Anti-Dixin Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1: 64000; Western Blot: 0.5-2?g/ml

Type: Primary
Antigen: Dixin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR