Anti-DPYSL2 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3575-55CEA 4320 DKK
USBID3575-55C
Anti-DPYSL2 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-1:1000; Immunofluorescence: 1:100-1:500; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: DPYSL2 (dihydropyrimidinase-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR