Anti-Dihydroorotate Dehydrogenase Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 10B2725]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3574-75-HRPEA 0 DKK
USBID3574-75-HRP
Anti-Dihydroorotate Dehydrogenase Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 10B2725]
Type: Primary
Antigen: Dihydroorotate Dehydrogenase
Clonality: monoclonal
Clone: 10B2725
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR