Anti-DICER1 Rabbit polyclonal antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBID3520-01TEA 3360 DKK
USBID3520-01T
Anti-DICER1 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 0.5-4?g/ml
Type: Primary
Antigen: DICER1 (dicer 1, ribonuclease type III)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat