Anti-DICER1 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3520-01REA 4030 DKK
USBID3520-01R
Anti-DICER1 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:200; Flow Cytometry: 1:100-1:1000

Type: Primary
Antigen: DICER1 (dicer 1, ribonuclease type III)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR