Anti-DICER1 Rabbit monoclonal antibody [clone: 11C1026]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3520-01GEA 4640 DKK
USBID3520-01G
Anti-DICER1 Rabbit monoclonal antibody [clone: 11C1026]
Antistoffer
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50

Type: Primary
Antigen: DICER1 (dicer 1, ribonuclease type III)
Clonality: monoclonal
Clone: 11C1026
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR