Anti-DBI Mouse monoclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3475-02BEA 4730 DKK
USBID3475-02B
Anti-DBI Mouse monoclonal antibody
Antistoffer
Recommended dilution: Immunochemistry: 1:200

Type: Primary
Antigen: DBI (diazepam binding inhibitor (GABA receptor modulator, acyl-CoA binding protein))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR