Anti-DBI Mouse monoclonal antibody [clone: 10K186]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3475-02AEA 3270 DKK
USBID3475-02A
Anti-DBI Mouse monoclonal antibody [clone: 10K186]
Antistoffer
Recommended dilution: Western Blot: 1:100-500; Immunoprecipitation: 1-2?g/ml s

Type: Primary
Antigen: DBI (diazepam binding inhibitor (GABA receptor modulator, acyl-CoA binding protein))
Clonality: monoclonal
Clone: 10K186
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR