Anti-DIAPH3 Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3470-30BEA 2750 DKK
USBID3470-30B
Anti-DIAPH3 Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:64000

Type: Primary
Antigen: DIAPH3 (diaphanous-related formin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR