Anti-DIAPH2 Mouse monoclonal antibody [clone: 12K79]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3470-23EA 4320 DKK
USBID3470-23
Anti-DIAPH2 Mouse monoclonal antibody [clone: 12K79]
Antistoffer
Recommended dilution: Western Blot: 2?g/ml

Type: Primary
Antigen: DIAPH2 (diaphanous-related formin 2)
Clonality: monoclonal
Clone: 12K79
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR