Anti-DMBT1 Rat Monoclonal Antibody [clone: 11n61]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3449-59CEA 4320 DKK
USBID3449-59C
Anti-DMBT1 Rat Monoclonal Antibody [clone: 11n61]
Antistoffer
Recommended dilution: Western Blot: 2?g/ml

Type: Primary
Antigen: DMBT1 (deleted in malignant brain tumors 1)
Clonality: monoclonal
Clone: 11n61
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR