Anti-DGKQ Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3441-88-PEEA 4620 DKK
USBID3441-88-PE
Anti-DGKQ Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antistoffer
Recommended dilution: FLISA: 1:1000; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: DGKQ (diacylglycerol kinase, theta 110kDa)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR