Anti-DGKB Mouse monoclonal antibody [clone: 11n59]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3441-82BEA 4320 DKK
USBID3441-82B
Anti-DGKB Mouse monoclonal antibody [clone: 11n59]
Antistoffer
Recommended dilution: Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: DGKB (diacylglycerol kinase, beta 90kDa)
Clonality: monoclonal
Clone: 11n59
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR