Anti-DAGLA Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3441-70EA 4320 DKK
USBID3441-70
Anti-DAGLA Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:128000; Western Blot: 0. 3-1 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: DAGLA (diacylglycerol lipase, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR