Anti-Diamine Oxidase Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3375-04-20EA 3500 DKK
USBID3375-04-20
Anti-Diamine Oxidase Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:4000; Western Blot: 1:1000-1:2000

Type: Primary
Antigen: Diamine Oxidase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR