Anti-DHEA-S Sheep polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3227-02EA 6850 DKK
USBID3227-02
Anti-DHEA-S Sheep polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1. 25 ?g/ml

Type: Primary
Antigen: DHEA-S
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR