Anti-DHEA Sheep polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3224-98EEA 2730 DKK
USBID3224-98E
Anti-DHEA Sheep polyclonal antibody
Antistoffer
Type: Primary
Antigen: DHEA
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR