Anti-DHEA Mouse monoclonal antibody [clone: 10B2718]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3224-98CEA 3970 DKK
USBID3224-98C
Anti-DHEA Mouse monoclonal antibody [clone: 10B2718]
Antistoffer
  • Recommended dilution: ELISA: 1:1000-1:10000

Type: Primary
Antigen: DHEA
Clonality: monoclonal
Clone: 10B2718
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR