Anti-DGAT2 Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3224-71CEA 4320 DKK
USBID3224-71C
Anti-DGAT2 Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:128000; Western Blot: 0. 3-1 ?g/ml; Immunochemistry: 3. 75 ?g/ml

Type: Primary
Antigen: DGAT2 (diacylglycerol O-acyltransferase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR