Anti-DFFB Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3224-30EEA 3210 DKK
USBID3224-30E
Anti-DFFB Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 0.5-4?g/ml; Immunochemistry: 2-8?g/ml

Type: Primary
Antigen: DFFB (DNA fragmentation factor, 40kDa, beta polypeptide (caspase-activated DNase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR