Anti-Developmental Pluripotency-Associated Protein 5 Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3221-94TEA 5650 DKK
USBID3221-94T
Anti-Developmental Pluripotency-Associated Protein 5 Goat polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 0.1-0.2?g/ml

Type: Primary
Antigen: Developmental Pluripotency-Associated Protein 5
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR