Anti-Desmoplakin 1 Mouse monoclonal antibody [clone: 5F191]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3221-89KEA 4870 DKK
USBID3221-89K
Anti-Desmoplakin 1 Mouse monoclonal antibody [clone: 5F191]
Antistoffer
Recommended dilution: Immunochemistry: 1:10

Type: Primary
Antigen: Desmoplakin 1
Clonality: monoclonal
Clone: 5F191
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Chicken, Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR