Anti-Desmoplakin 1 Guinea pig polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3221-89JEA 4760 DKK
USBID3221-89J
Anti-Desmoplakin 1 Guinea pig polyclonal antibody
Antistoffer
Recommended dilution: Immunochemistry: 1:100; Western Blot: 1:1000

Type: Primary
Antigen: Desmoplakin 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Guinea pig
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR