Anti-DSG4 Sheep Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3221-85EA 5920 DKK
USBID3221-85
Anti-DSG4 Sheep Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 0.1 ?g/ml

Type: Primary
Antigen: DSG4 (desmoglein 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR