Anti-DSG3 Mouse Monoclonal Antibody [clone: 3G133]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3221-84-50EA 2680 DKK
USBID3221-84-50 USBID3221-84-100
Anti-DSG3 Mouse Monoclonal Antibody [clone: 3G133]
Antistoffer
Type: Primary
Antigen: DSG3 (desmoglein 3)
Clonality: monoclonal
Clone: 3G133
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR