Anti-DSG2 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3221-83GEA 4380 DKK
USBID3221-83G
Anti-DSG2 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:50-1:100; ELISA: 1:1000; Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: DSG2 (desmoglein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR