Anti-DSG2 Mouse Monoclonal Antibody [clone: 6A539]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3221-83CEA 4410 DKK
USBID3221-83C
Anti-DSG2 Mouse Monoclonal Antibody [clone: 6A539]
Antistoffer
Recommended dilution: Immunofluorescence: 1:10; Western Blot: 1:10

Type: Primary
Antigen: DSG2 (desmoglein 2)
Clonality: monoclonal
Clone: 6A539
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR