Anti-DSG1 Mouse Monoclonal Antibody [clone: 27B2]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3221-82DEA 6690 DKK
USBID3221-82D
Anti-DSG1 Mouse Monoclonal Antibody [clone: 27B2]
Antistoffer
Recommended dilution: Immunofluorescence: 1:10; Western Blot: 1:10

Type: Primary
Antigen: DSG1 (desmoglein 1)
Clonality: monoclonal
Clone: 27B2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR