Anti-DSC3 Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10C15]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3221-52C-APEA 0 DKK
USBID3221-52C-AP
Anti-DSC3 Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10C15]
Type: Primary
Antigen: DSC3 (desmocollin 3)
Clonality: monoclonal
Clone: 10C15
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR