Anti-DES Mouse monoclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3221-10KEA 4320 DKK
USBID3221-10K
Anti-DES Mouse monoclonal antibody
Antistoffer
  • Recommended dilution: ELISA: 1:10000; Western Blot: 1:1000-1:2000; Immunochemistry: 1:1000-1:2000;

Type: Primary
Antigen: DES (desmin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR