Anti-DES Mouse monoclonal antibody [clone: 5F179]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3221-06AEA 3460 DKK
USBID3221-06A
Anti-DES Mouse monoclonal antibody [clone: 5F179]
Antistoffer
Recommended dilution: Immunochemistry: 1:10-1:20

Type: Primary
Antigen: DES (desmin)
Clonality: monoclonal
Clone: 5F179
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mammalian, Mouse, Rabbit, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR