Anti-DES Mouse monoclonal antibody [clone: 37EH]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3221-04DEA 4090 DKK
USBID3221-04D
Anti-DES Mouse monoclonal antibody [clone: 37EH]
Antistoffer
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: DES (desmin)
Clonality: monoclonal
Clone: 37EH
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR