Anti-DES Mouse monoclonal antibody [clone: 9L152]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3221-03CEA 2940 DKK
USBID3221-03C
Anti-DES Mouse monoclonal antibody [clone: 9L152]
Antistoffer
Type: Primary
Antigen: DES (desmin)
Clonality: monoclonal
Clone: 9L152
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR