Anti-DES Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3221-02DEA 4320 DKK
USBID3221-02D
Anti-DES Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:2000; Western Blot: 0.1-0. 3?g/ml; Immunochemistry: 3. 75 ?g/ml

Type: Primary
Antigen: DES (desmin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR